accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CCH3NHOHO

CAS RN 318-98-9

IUPAC Name: hydrogen 1-(naphthalen-1-yloxy)-3-[(propan-2-yl)amino]propan-2-ol chloride
Synonyms: propranolol hydrochloride
Molecular Weight (g/mol): 295.81
Molecular Formula: C16H22ClNO2
InChi Key: ZMRUPTIKESYGQW-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)NCC(O)COC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (8)
Percent Purity 
  • (2)
  • (8)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (8)
  • (3)
Grade 
  • (1)
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (8)
  • (1)
  • (8)
  • (2)
  • (5)
  • (8)
  • (3)
  • (4)
  • (8)
  • (2)
  • (8)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (8)
  • (3)
  • (1)
  • (8)