Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propranolol Hydrochloride, USP, 98-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18605895 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
2.5 kg
25 kg
Packaging:
Amber Glass Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-895 500 g Amber Glass Bottle
18-605-888 1 g Amber Glass Bottle
18-605-889 5 g Amber Glass Bottle
18-605-890 25 g Amber Glass Bottle
18-605-891 100 g Amber Glass Bottle
18-605-892 1 kg Amber Glass Bottle
18-605-893 2.5 kg Amber Glass Bottle
18-605-894 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-605-895 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PR140500GMBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PR140, 318-98-9, C16H21NO2·HCl

Spectrum™ Chemical Propranolol Hydrochloride, USP is a sympatholytic nonselective beta blocker used to treat anxiety, hypertension and panic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 318-98-9
Molecular Formula C16H22ClNO2
Molecular Weight (g/mol) 295.81
InChI Key ZMRUPTIKESYGQW-UHFFFAOYNA-N
IUPAC Name hydrogen 1-(naphthalen-1-yloxy)-3-[(propan-2-yl)amino]propan-2-ol chloride
SMILES [H+].[Cl-].CC(C)NCC(O)COC1=C2C=CC=CC2=CC=C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 162-165°C
Packaging Amber Glass Bottle
Quantity 500 g
Specific Rotation -1.0° to +1.0°
Percent Purity 98 to 101.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Propranolol Hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.