accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3188-13-4

IUPAC Name: (chloromethoxy)ethane
Synonyms: chloromethyl ethyl ether
Molecular Weight (g/mol): 94.54
Molecular Formula: C3H7ClO
InChi Key: FCYRSDMGOLYDHL-UHFFFAOYSA-N
SMILES: CCOCCl
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)