accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHH2OClClCH3NNCl

CAS RN 31884-77-2

IUPAC Name: dihydrogen 1-[(4-chlorophenyl)(phenyl)methyl]-4-[(3-methylphenyl)methyl]piperazine hydrate dichloride
Synonyms: dihydrogen meclizine hydrate dichloride
Molecular Weight (g/mol): 481.89
Molecular Formula: C25H31Cl3N2O
InChi Key: KDLHYOMCWBWLMM-UHFFFAOYNA-N
SMILES: [H+].[H+].O.[Cl-].[Cl-].CC1=CC=CC(CN2CCN(CC2)C(C2=CC=CC=C2)C2=CC=C(Cl)C=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results