Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meclizine dihydrochloride monohydrate, Thermo Scientific™

Catalog No. AAJ6246406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6246406 5 g
AAJ6246403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6246406 Supplier Thermo Scientific Chemicals Supplier No. J6246406

Antiemetic and H1-histamine receptor antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Antiemetic and H1-histamine receptor antagonistMeclizine dihydrochloride monohydrate acts as an antiemetic. It is also used for the prevention and treatment of nausea during pregnancy. In addition to this, it is used in radiation sickness, vomiting or dizziness associated with motion sickness.

Solubility
Insoluble in water.

Notes
Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31884-77-2
Molecular Formula C25H31Cl3N2O
Molecular Weight (g/mol) 481.886
MDL Number MFCD00058199
InChI Key KDLHYOMCWBWLMM-UHFFFAOYSA-N
Synonym meclizine hydrochloride, meclizine dihydrochloride monohydrate, meclizine hcl, antivert tn, c25h27cln2.2hcl.h2o, 1-p-chloro-alpha-phenylbenzyl-4-m-methylbenzyl piperazine dihydrochloride monohydrate, 1-4-chlorophenyl-phenylmethyl-4-3-methylphenyl methyl piperazine hydrate dihydrochloride, piperazine, 1-4-chlorophenyl phenylmethyl-4-3-methylphenyl methyl-, dihydrochloride, monohydrate, meclizine hcl hydrate, meclizine hydrochloride usan:jan
PubChem CID 173612
IUPAC Name 1-[(4-chlorophenyl)-phenylmethyl]-4-[(3-methylphenyl)methyl]piperazine;hydrate;dihydrochloride
SMILES CC1=CC(=CC=C1)CN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=C(C=C4)Cl.O.Cl.Cl

Specifications

Melting Point 210°C to 213°C
Color White
Quantity 5 g
Merck Index 14,5777
Solubility Information Insoluble in water.
Formula Weight 481.89 (463.88 Anhydrous)
Physical Form Powder
Chemical Name or Material Meclizine dihydrochloride monohydrate
Hazard Category H302-H361
Hazard Statement GHS H Statement
H361-H302
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
TSCA Yes
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.