accessibility menu, dialog, popup

UserName

Viewing profile as user:
Fe(R)CH3NCH3H3C

CAS RN 31886-58-5

IUPAC Name: (R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine
Synonyms: —
Molecular Weight (g/mol): 257.16
Molecular Formula: C14H19FeN
InChi Key: —
SMILES: [Fe].c1cccc1.C[C@@H](N(C)C)c1cccc1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)