Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine, 97%, Thermo Scientific Chemicals

Catalog No. AAL19679MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19679MD 250 mg
AAL1967903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19679MD Supplier Thermo Scientific Chemicals Supplier No. L19679MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31886-58-5
Molecular Formula C14H19FeN
Molecular Weight (g/mol) 257.16
MDL Number MFCD00066273
IUPAC Name (R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine
SMILES [Fe].c1cccc1.C[C@@H](N(C)C)c1cccc1

Specifications

Density 1.222
Boiling Point 120°C to 121°C (0.7 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.589
Quantity 250 mg
Optical Rotation +13° (c=1 in Ethanol, 5H)
Formula Weight 257.16
Percent Purity 97%
Chemical Name or Material (R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.