accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONNN

CAS RN 3189-43-3

IUPAC Name: oxirane-2,2,3,3-tetracarbonitrile
Synonyms: oxirane-2,2,3,3-tetracarbonitrile
Molecular Weight (g/mol): 144.09
Molecular Formula: C6N4O
InChi Key: YWXLYZIZWVOMML-UHFFFAOYSA-N
SMILES: N#CC1(OC1(C#N)C#N)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results