Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracyanoethylene Oxide 97.0+%, TCI America™
SDP

Catalog No. T14391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T14391G Supplier TCI America Supplier No. T14391G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3189-43-3
Molecular Formula C6N4O
Molecular Weight (g/mol) 144.093
MDL Number MFCD00014483
InChI Key YWXLYZIZWVOMML-UHFFFAOYSA-N
Synonym Oxiranetetracarbonitrile
PubChem CID 76661
IUPAC Name oxirane-2,2,3,3-tetracarbonitrile
SMILES C(#N)C1(C(O1)(C#N)C#N)C#N

Specifications

Melting Point 175°C
Quantity 1 g
UN Number 3439
Formula Weight 144.09
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tetracyanoethylene Oxide
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.