accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClBrI

CAS RN 31928-44-6

IUPAC Name: 2-bromo-4-chloro-1-iodobenzene
Synonyms: 2-bromo-4-chloro-1-iodobenzene
Molecular Weight (g/mol): 317.35
Molecular Formula: C6H3BrClI
InChi Key: CXHXFDQEFKFYQJ-UHFFFAOYSA-N
SMILES: ClC1=CC(Br)=C(I)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)