Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-chloro-1-iodobenzene, 98%

Catalog No. AC448760250
Change view
Click to view available options
Quantity:
1 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC448760250 25 g Glass bottle
AC448760010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448760250 Supplier Thermo Scientific Chemicals Supplier No. 448760250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 31928-44-6 | C6H3BrClI | 317.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31928-44-6
Molecular Formula C6H3BrClI
Molecular Weight (g/mol) 317.36
InChI Key CXHXFDQEFKFYQJ-UHFFFAOYSA-N
Synonym 2-bromo-4-chloroiodobenzene, benzene, 2-bromo-4-chloro-1-iodo, acmc-209hpo, ksc494q6f, 1-iodo-2-bromo-4-chlorobenzene, 2-bromo-1-iodo-4-chlorobenzene
PubChem CID 13741326
IUPAC Name 2-bromo-4-chloro-1-iodobenzene
SMILES C1=CC(=C(C=C1Cl)Br)I

Specifications

Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(Cl)I
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 317.36
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-chloro-1-iodobenzene
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.