accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31930-34-4

IUPAC Name: ethyl (2E)-3-bromoprop-2-enoate
Synonyms: ethyl (2E)-3-bromoprop-2-enoate
Molecular Weight (g/mol): 179.01
Molecular Formula: C5H7BrO2
InChi Key: UJTJVQIYRQALIK-ONEGZZNKSA-N
SMILES: CCOC(=O)\C=C\Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results