accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31930-36-6

IUPAC Name: ethyl 3-iodoprop-2-enoate
Synonyms: ethyl 3-iodoprop-2-enoate
Molecular Weight (g/mol): 226.01
Molecular Formula: C5H7IO2
InChi Key: AELYFQSZXFFNGP-UHFFFAOYSA-N
SMILES: CCOC(=O)C=CI
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)