accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CNNNH2NNH2

CAS RN 3194-70-5

IUPAC Name: 6-ethenyl-1,3,5-triazine-2,4-diamine
Synonyms: 6-ethenyl-1,3,5-triazine-2,4-diamine
Molecular Weight (g/mol): 137.15
Molecular Formula: C5H7N5
InChi Key: ZXLYUNPVVODNRE-UHFFFAOYSA-N
SMILES: NC1=NC(C=C)=NC(N)=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results