Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Vinyl-4,6-diamino-1,3,5-triazine 95.0+%, TCI America™
SDP

Catalog No. V00571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V00571G Supplier TCI America Supplier No. V00571G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3194-70-5
Molecular Formula C5H7N5
Molecular Weight (g/mol) 137.15
MDL Number MFCD00059935
InChI Key ZXLYUNPVVODNRE-UHFFFAOYSA-N
Synonym 4,6-Diamino-2-vinyl-1,3,5-triazine
PubChem CID 586156
IUPAC Name 6-ethenyl-1,3,5-triazine-2,4-diamine
SMILES NC1=NC(C=C)=NC(N)=N1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 137.15
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Vinyl-4,6-diamino-1,3,5-triazine
EINECSNumber (5)-6522
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.