accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNCH2FFFSOONSOOFFF

CAS RN 319476-28-3

IUPAC Name: 1-ethenyl-3-ethyl-1H-imidazol-3-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: 3-ethenyl-1-ethylimidazol-1-ium; bistriflylimide anion
Molecular Weight (g/mol): 403.31
Molecular Formula: C9H11F6N3O4S2
InChi Key: DFGRLSLTNVNMEM-UHFFFAOYSA-N
SMILES: CC[N+]1=CN(C=C)C=C1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results