Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethyl-1-vinylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
SDP

Catalog No. E12805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E12805G Supplier TCI America Supplier No. E12805G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 319476-28-3
Molecular Formula C9H11F6N3O4S2
Molecular Weight (g/mol) 403.314
InChI Key DFGRLSLTNVNMEM-UHFFFAOYSA-N
PubChem CID 121235111
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1-ethenyl-3-ethylimidazol-3-ium
SMILES CC[N+]1=CN(C=C1)C=C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point 18°C
Color Yellow
Quantity 5 g
Formula Weight 403.31
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 3-Ethyl-1-vinylimidazolium Bis(trifluoromethanesulfonyl)imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.