CAS RN 32017-76-8
IUPAC Name:
2-(4-bromophenyl)oxirane
Synonyms:
2-(4-bromophenyl)oxirane
Molecular Weight (g/mol):
199.05
Molecular Formula:
C8H7BrO
InChi Key:
NNINSLOEPXEZOZ-UHFFFAOYNA-N
SMILES:
BrC1=CC=C(C=C1)C1CO1
1
–
1
of
1
results
1
Sigma Aldrich 2-(4-Bromophenyl)oxirane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.