accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32083-66-2

IUPAC Name: 2-(4-fluorobenzenesulfonyl)acetonitrile
Synonyms: 2-(4-fluorobenzenesulfonyl)acetonitrile
Molecular Weight (g/mol): 199.2
Molecular Formula: C8H6FNO2S
InChi Key: WBXJZTLPEMITJL-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)S(=O)(=O)CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results