accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 321-64-2

ClNH3N

CAS RN 321-64-2

IUPAC Name: 1,2,3,4-tetrahydroacridin-9-aminium chloride
Synonyms: tacrine
Molecular Weight (g/mol): 234.73
Molecular Formula: C13H15ClN2
InChi Key: ZUFVXZVXEJHHBN-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]C1=C2C=CC=CC2=NC2=C1CCCC2
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results