Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Amino-1,2,3,4-tetrahydroacridine Hydrochloride Hydrate 99+%, Thermo Scientific™

Catalog No. AC133500050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC133500050 5 g
AC133500010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC133500050 Supplier Thermo Scientific Chemicals Supplier No. 133500050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 321-64-2
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
InChI Key YLJREFDVOIBQDA-UHFFFAOYSA-N
Synonym tacrine, tetrahydroaminacrine, tetrahydroaminoacridine, 1,2,3,4-tetrahydro-9-acridinamine, cognex, tetrahydroaminocrin, tetrahydroaminocrine, 9-amino-1,2,3,4-tetrahydroacridine, romotal, tacrinum
PubChem CID 1935
ChEBI CHEBI:45980
IUPAC Name 1,2,3,4-tetrahydroacridin-9-amine
SMILES C1CCC2=NC3=CC=CC=C3C(=C2C1)N

Specifications

Quantity 5 g
Beilstein 22, III, 4911
Merck Index 14, 9024
Formula Weight 234.72
Chemical Name or Material 9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate, 99+%
EINECSNumber 206-291-2
RTECSNumber AR9532100
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.