accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 321182-37-0

FFH2NFNH2

CAS RN 321182-37-0

IUPAC Name: 2,4,5-trifluorobenzene-1,3-diamine
Synonyms: 2,4,5-trifluorobenzene-1,3-diamine
Molecular Weight (g/mol): 162.12
Molecular Formula: C6H5F3N2
InChi Key: ONVSXVVJFJJZQX-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(F)C(N)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results