accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32133-82-7

IUPAC Name: [2-({diphenyl[(1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-yl)oxy]-λ⁴-sulfanyl}oxy)-1,1,1,3,3,3-hexafluoropropan-2-yl]benzene
Synonyms: [2-({diphenyl[(1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-yl)oxy]-λ⁴-sulfanyl}oxy)-1,1,1,3,3,3-hexafluoropropan-2-yl]benzene
Molecular Weight (g/mol): 672.53
Molecular Formula: C30H20F12O2S
InChi Key: RMIBJVUYNZSLSD-UHFFFAOYSA-N
SMILES: FC(F)(F)C(OS(OC(C1=CC=CC=C1)(C(F)(F)F)C(F)(F)F)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results