Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis[alpha,alpha-bis(trifluoromethyl)benzenemethanolato]diphenylsulfur 96.0+%, TCI America™
SDP

Catalog No. B22005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22005G 5 g
B22001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22005G Supplier TCI America Supplier No. B22005G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32133-82-7
Molecular Formula C30H20F12O2S
Molecular Weight (g/mol) 672.53
MDL Number MFCD00010662
InChI Key RMIBJVUYNZSLSD-UHFFFAOYSA-N
Synonym Martin Sulfurane
PubChem CID 3608068
IUPAC Name [2-({diphenyl[(1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-yl)oxy]-λ⁴-sulfanyl}oxy)-1,1,1,3,3,3-hexafluoropropan-2-yl]benzene
SMILES FC(F)(F)C(OS(OC(C1=CC=CC=C1)(C(F)(F)F)C(F)(F)F)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C(F)(F)F

Specifications

Melting Point 111°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 672.53
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis[alpha,alpha-bis(trifluoromethyl)benzenemethanolato]diphenylsulfur
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.