accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3215-64-3

IUPAC Name: 2-(2,6-dichlorophenyl)acetonitrile
Synonyms: 2-(2,6-dichlorophenyl)acetonitrile
Molecular Weight (g/mol): 186.04
Molecular Formula: C8H5Cl2N
InChi Key: AOEJUUCUKRUCEF-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1CC#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results