Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorobenzyl Cyanide 98.0+%, TCI America™
SDP

Catalog No. D179325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D179325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D179325G Supplier TCI America Supplier No. D179325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3215-64-3
Molecular Formula C8H5Cl2N
Molecular Weight (g/mol) 186.035
MDL Number MFCD00001901
InChI Key AOEJUUCUKRUCEF-UHFFFAOYSA-N
Synonym 2,6-dichlorophenylacetonitrile, 2,6-dichlorobenzyl cyanide, 2-2,6-dichlorophenyl acetonitrile, 2,6-dichlorophenyl acetonitrile, 2,6-dichlorobenzeneacetonitrile, benzeneacetonitrile, 2,6-dichloro, acetonitrile, 2,6-dichlorophenyl, 2,6-dichloro-phenyl-acetonitrile, 2-2,6-dichlorophenyl ethanenitrile
PubChem CID 76678
IUPAC Name 2-(2,6-dichlorophenyl)acetonitrile
SMILES C1=CC(=C(C(=C1)Cl)CC#N)Cl

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 186.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorobenzyl Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.