accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2On

CAS RN 32171-39-4

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C2H4O)nC4H6O2
InChi Key: HFCUBKYHMMPGBY-UHFFFAOYSA-N
SMILES: COCCOC(=O)C=C
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)