Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) methyl ether acrylate

Catalog No. 454990250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-499-0250ML 250 mL
45-499-01L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-499-0250ML Supplier Sigma Aldrich Supplier No. 454990250ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(ethylene glycol) methyl ether methacrylate (PEGMA) is a nonlinear analog of polyethylene glycol (PEG). It is a biocompatible homopolymer with a brush type structure that is mainly used to provide a PEG modified surface.PEGMA can be used in the development of molecular brushes as a substrate material for stimuli-responsive surfaces in biomedical applications. It may be used in the surface modification of poly(ether sulfone) based ultrafiltration (UF) membrane as a foul-resistant material.

Specifications

CAS 32171-39-4
Density 1.09 g/mL (at 25°C)
Linear Formula H2C=CHCO2(CH2CH2O)nCH3
MDL Number MFCD00803685
Quantity 250 mL
Synonym Acryl-PEG; Methoxy PEG acrylate; Methoxy poly(ethylene glycol) monoacrylate; Poly(ethylene glycol) monomethyl ether monoacrylate; mPEG-acrylate
Molecular Weight (g/mol) 480
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.