accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 3218-36-8

IUPAC Name: [1,1'-biphenyl]-4-carbaldehyde
Synonyms: [1,1'-biphenyl]-4-carbaldehyde
Molecular Weight (g/mol): 182.22
Molecular Formula: C13H10O
InChi Key: ISDBWOPVZKNQDW-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (3)
  • (4)
  • (1)
  • (4)
Boiling Point 
  • (3)
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (6)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (6)
  • (1)
  • (3)
  • (4)
  • (1)
  • (4)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)