Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl-4-carboxaldehyde,MP Biomedicals

Catalog No. ICN216591 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN216591 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN216591 Supplier MP Biomedicals, Inc Supplier No. 0521659105

Chemical Identifiers

CAS 3218-36-8
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.22
MDL Number MFCD00006947
InChI Key ISDBWOPVZKNQDW-UHFFFAOYSA-N
Synonym 4-biphenylcarboxaldehyde, 4-biphenylaldehyde, p-phenylbenzaldehyde, 1,1'-biphenyl-4-carbaldehyde, biphenyl-4-carboxaldehyde, 4-formylbiphenyl, p-biphenylylaldehyde, 1,1'-biphenyl-4-carboxaldehyde, p-biphenylaldehyde, p-biphenylcarboxaldehyde
PubChem CID 76689
IUPAC Name [1,1'-biphenyl]-4-carbaldehyde
SMILES O=CC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Quantity 10 g
Formula Weight 182.22
Chemical Name or Material Diphenyl-4-Carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.