accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3CHO

CAS RN 3219-63-4

IUPAC Name: (trimethylsilyl)methanol
Synonyms: (trimethylsilyl)methanol
Molecular Weight (g/mol): 104.22
Molecular Formula: C4H12OSi
InChi Key: ZQKNBDOVPOZPLY-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CO
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)