accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 321921-71-5

CH3P

CAS RN 321921-71-5

IUPAC Name: bis(adamantan-1-yl)(butyl)phosphane
Synonyms: bis(adamantan-1-yl)(butyl)phosphane
Molecular Weight (g/mol): 358.55
Molecular Formula: C24H39P
InChi Key: HTJWUNNIRKDDIV-UHFFFAOYSA-N
SMILES: CCCCP(C12CC3CC(CC(C3)C1)C2)C12CC3CC(CC(C3)C1)C2
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)