Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Catacxium™ A

Catalog No. 6714795G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
100 g
250 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-147-95G 5 g
67-147-950G 50 g
67-147-91G 1 g
67-147-9100G 100 g
67-147-9250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 67-147-95G Supplier Sigma Aldrich Supplier No. 6714795G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sold in collaboration with Solvias AGcataCXium™ A or di-adamantylalkylphosphine is a bulky and electron-rich phosphine ligand that is highly effective for palladium catalyzed cross-coupling reactions such as Heck and Suzuki coupling, Buchwald-Hartwig amination of aryl chlorides, and +a-arylation reactions of ketones. Other applications:palladium-catalyzed carbonylation of aryl and heteroaryl halidespalladium-catalyzed synthesis of (hetero)aromatic nitrilespalladium-catalyzed aminocarbonylation of aryl halidescataCXium is a registered trademark of Evonik Degussa GmbH

Specifications

CAS 321921-71-5
Molecular Formula C24H39P
Linear Formula C24H39P
MDL Number MFCD05861606
Quantity 5 g
Synonym Di(1-adamantyl)-n-butylphosphine
Molecular Weight (g/mol) 358.54 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.