accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 322-46-3

IUPAC Name: pyrido[2,3-b]pyrazine
Synonyms: pyrido(2,3)pyrazine
Molecular Weight (g/mol): 131.14
Molecular Formula: C7H5N3
InChi Key: YEYHFKBVNARCNE-UHFFFAOYSA-N
SMILES: C1=CN=C2N=CC=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results