accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHOCH3

CAS RN 32233-43-5

IUPAC Name: 2-(2,2-dimethyl-1,3-dioxolan-4-yl)ethan-1-ol
Synonyms: 2-(2,2-dimethyl-1,3-dioxolan-4-yl)ethanol
Molecular Weight (g/mol): 146.19
Molecular Formula: C7H14O3
InChi Key: YYEZYENJAMOWHW-UHFFFAOYNA-N
SMILES: CC1(C)OCC(CCO)O1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)