Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-4-(2-Hydroxyethyl)-2,2-dimethyl-1,3-dioxolane 95.0+%, TCI America™
SDP

Catalog No. H11891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H11891G Supplier TCI America Supplier No. H11891G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32233-43-5
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.186
MDL Number MFCD02682967
InChI Key YYEZYENJAMOWHW-ZCFIWIBFSA-N
Synonym (S)-2-(2,2-Dimethyl-1,3-dioxolan-4-yl)ethanol
PubChem CID 11480522
IUPAC Name 2-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethanol
SMILES CC1(OCC(O1)CCO)C

Specifications

Color Yellow
Boiling Point 92°C
Quantity 1 g
Formula Weight 146.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-4-(2-Hydroxyethyl)-2,2-dimethyl-1,3-dioxolane
EINECSNumber (5)-0502
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.