accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 322413-12-7

IUPAC Name: 1-(4-methoxyphenyl)-N-[4-(2-phenylethenyl)phenyl]methanimine
Synonyms: 1-(4-methoxyphenyl)-N-[4-(2-phenylethenyl)phenyl]methanimine
Molecular Weight (g/mol): 313.4
Molecular Formula: C22H19NO
InChi Key: AMWLHYSTQXHJRQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=NC2=CC=C(C=CC3=CC=CC=C3)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results