Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[(Methoxybenzylidene)amino]stilbene 98.0+%, TCI America™
SDP

Catalog No. M06031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M06031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M06031G Supplier TCI America Supplier No. M06031G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 322413-12-7
Molecular Formula C22H19NO
Molecular Weight (g/mol) 313.40
MDL Number MFCD00059296
InChI Key AMWLHYSTQXHJRQ-UHFFFAOYSA-N
Synonym 4-(p-Anisalamino)stilbene, 4-[(p-Anisylidene)amino]stilbene
PubChem CID 5378812
IUPAC Name 1-(4-methoxyphenyl)-N-[4-(2-phenylethenyl)phenyl]methanimine
SMILES COC1=CC=C(C=NC2=CC=C(C=CC3=CC=CC=C3)C=C2)C=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 313.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[(Methoxybenzylidene)amino]stilbene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.