CAS RN 32249-35-7
IUPAC Name:
methyl 3-cyclopropyl-3-oxopropanoate
Synonyms:
methyl 3-cyclopropyl-3-oxopropanoate
Molecular Weight (g/mol):
142.15
Molecular Formula:
C7H10O3
InChi Key:
RIJWDPRXCXJDPK-UHFFFAOYSA-N
SMILES:
COC(=O)CC(=O)C1CC1
1
–
1
of
1
results
1
Methyl 3-Cyclopropyl-3-oxopropionate 96.0+%, TCI America™
CAS: 32249-35-7 Molecular Formula: C7H10O3 Molecular Weight (g/mol): 142.15 MDL Number: MFCD00274090 InChI Key: RIJWDPRXCXJDPK-UHFFFAOYSA-N Synonym: 3-Cyclopropyl-3-oxopropionic Acid Methyl Ester PubChem CID: 10130016 IUPAC Name: methyl 3-cyclopropyl-3-oxopropanoate SMILES: COC(=O)CC(=O)C1CC1