accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3225-74-9

IUPAC Name: 2-amino-1-(2-fluorophenyl)ethan-1-ol
Synonyms: 2-amino-1-(2-fluorophenyl)ethanol
Molecular Weight (g/mol): 155.17
Molecular Formula: C8H10FNO
InChi Key: MFGOFGRYDNHJTA-UHFFFAOYNA-N
SMILES: NCC(O)C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results