Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-1-(2-fluorophenyl)ethanol, 99%, Thermo Scientific™

Catalog No. AAH50770MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50770MD 250 mg
AAH5077003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50770MD Supplier Thermo Scientific Chemicals Supplier No. H50770MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Amino-1-(2-fluorophenyl)ethanol is used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3225-74-9
Molecular Formula C8H10FNO
Molecular Weight (g/mol) 155.172
MDL Number MFCD08060976
InChI Key MFGOFGRYDNHJTA-UHFFFAOYSA-N
Synonym 2-amino-1-2-fluorophenyl ethanol, 2-amino-1-2-fluoro-phenyl-ethanol, 2-amino-1-2-fluorophenyl ethan-1-ol, acmc-1aewg, 2-fluorophenylethanolamine, 2-2-fluorophenyl-2-hydroxyethanamine, 2-hydroxy-2-2-fluorophenyl ethanamine, 2-hydroxy-2-2-fluorophenyl-ethanamine
PubChem CID 15067176
IUPAC Name 2-amino-1-(2-fluorophenyl)ethanol
SMILES C1=CC=C(C(=C1)C(CN)O)F

Specifications

Quantity 250 mg
Formula Weight 155.17
Percent Purity 99%
Chemical Name or Material 2-Amino-1-(2-fluorophenyl)ethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.