accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32270-93-2

IUPAC Name: dichlorocopper; bis(propane-1,3-diamine)
Synonyms: copper chloride; bis(α,ω-propanediamine)
Molecular Weight (g/mol): 282.7
Molecular Formula: C6H20Cl2CuN4
InChi Key: HSUSKWQWIPIFEB-UHFFFAOYSA-L
SMILES: Cl[Cu]Cl.NCCCN.NCCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results