Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(1,3-propanediamine) Copper(II) Dichloride 98.0+%, TCI America™
SDP

Catalog No. B15131G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15131G 1 g
B151310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15131G Supplier TCI America Supplier No. B15131G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32270-93-2
Molecular Formula C6H20Cl2CuN4
Molecular Weight (g/mol) 282.7
MDL Number MFCD00191318
InChI Key HSUSKWQWIPIFEB-UHFFFAOYSA-L
PubChem CID 12980278
IUPAC Name dichlorocopper;propane-1,3-diamine
SMILES C(CN)CN.C(CN)CN.Cl[Cu]Cl

Specifications

Color Blue-Purple
Quantity 1 g
Formula Weight 282.70
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(1,3-propanediamine) Copper(II) Dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.