accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3230-39-5

IUPAC Name: 4-{[(4-methoxyphenyl)methylidene]amino}phenol
Synonyms: 4-{[(4-methoxyphenyl)methylidene]amino}phenol
Molecular Weight (g/mol): 227.26
Molecular Formula: C14H13NO2
InChi Key: YONXPYGTYHMKDH-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=NC2=CC=C(O)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results