Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Methoxybenzylidene)-4-hydroxyaniline 98.0+%, TCI America™
SDP

Catalog No. M06055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M06055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M06055G Supplier TCI America Supplier No. M06055G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3230-39-5
Molecular Formula C14H13NO2
Molecular Weight (g/mol) 227.263
MDL Number MFCD00037176
InChI Key YONXPYGTYHMKDH-UHFFFAOYSA-N
Synonym N-(p-Anisal)-4-hydroxyaniline, N-(p-Anisylidene)-4-hydroxyaniline, 4-[(4-Methoxybenzylidene)amino]phenol
PubChem CID 97286
IUPAC Name 4-[(4-methoxyphenyl)methylideneamino]phenol
SMILES COC1=CC=C(C=C1)C=NC2=CC=C(C=C2)O

Specifications

Melting Point 192°C
Color Yellow
Quantity 5 g
Formula Weight 227.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Methoxybenzylidene)-4-hydroxyaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.