accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3H3CHC

CAS RN 3234-64-8

IUPAC Name: 3-ethylpent-1-yn-3-aminium
Synonyms: 3-ethylpent-1-yn-3-aminium
Molecular Weight (g/mol): 112.2
Molecular Formula: C7H14N
InChi Key: WHNOKDNCUNZBLW-UHFFFAOYSA-O
SMILES: CCC([NH3+])(CC)C#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)