Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Diethylpropargylamine, 97%, Thermo Scientific™

Catalog No. AC175635000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC175635000 500 mL
AC175631000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC175635000 Supplier Thermo Scientific Chemicals Supplier No. 175635000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3234-64-8
Molecular Formula C7H14N
Molecular Weight (g/mol) 112.20
MDL Number MFCD00008057
InChI Key WHNOKDNCUNZBLW-UHFFFAOYSA-O
Synonym 1,1-diethylpropargylamine, 1-pentyn-3-amine, 3-ethyl, 1-pentyn-3-amine,3-ethyl, 1,1-diethyl-prop-2-ynylamine, 2,2-diethylpropargylamine, 1,1-diethylproparagyl amine, 3-ethyl-1-pentyn-3-amine, 3-amino-3-ethyl-1-pentyne, 1,1-diethyl-2-propynylamine, 1,1-diethyl-2-propynylamine #
PubChem CID 76710
IUPAC Name 3-ethylpent-1-yn-3-aminium
SMILES CCC([NH3+])(CC)C#C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.82g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Refractive Index 1.4399 to 1.4419 (20°C, 589nm)
Linear Formula HC2CC(C2H5)2NH2
Quantity 500 mL
Formula Weight 111.19
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 1, 1-Diethylpropargylamine, 97%
Show More Show Less
EINECSNumber 221-784-2
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.