CAS RN 3236-63-3
IUPAC Name:
2-[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
Synonyms:
2-[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
Molecular Weight (g/mol):
228.29
Molecular Formula:
C15H16O2
InChi Key:
XZXYQEHISUMZAT-UHFFFAOYSA-N
SMILES:
CC1=CC=C(O)C(CC2=CC(C)=CC=C2O)=C1
1
–
1
of
1
results
1
2,2'-Methylenebis(4-methylphenol) 90.0+%, TCI America™
CAS: 3236-63-3 Molecular Formula: C15H16O2 Molecular Weight (g/mol): 228.29 MDL Number: MFCD00155176 InChI Key: XZXYQEHISUMZAT-UHFFFAOYSA-N PubChem CID: 76715 IUPAC Name: 2-[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol SMILES: CC1=CC=C(O)C(CC2=CC(C)=CC=C2O)=C1