Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Methylenebis(4-methylphenol) 90.0+%, TCI America™
SDP

Catalog No. M11345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11345G 5 g
M113425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11345G Supplier TCI America Supplier No. M11345G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3236-63-3
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.29
MDL Number MFCD00155176
InChI Key XZXYQEHISUMZAT-UHFFFAOYSA-N
PubChem CID 76715
IUPAC Name 2-[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
SMILES CC1=CC=C(O)C(CC2=CC(C)=CC=C2O)=C1

Specifications

Melting Point 125°C
Color White
Quantity 5 g
Synonym Bis(2-hydroxy-5-methylphenyl)methane, 2,2′C-Dihydroxy-5,5′C-dimethyldiphenylmethane, 2,2′C-Methylenebis(p-cresol)
Formula Weight 228.29
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Methylenebis(4-methylphenol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.