accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3238-55-9

IUPAC Name: 1-(pyridin-2-yl)propan-1-one
Synonyms: 1-(pyridin-2-yl)propan-1-one
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: ZHAZHKPVEROFLH-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results